Ammonium sulfate ≥99%, granular FCC, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Diammonium sulfate
, Sulphuric acid diammonium salt
, sulfuric acid diammonium salt
, Diammonium sulphate
, Mascagnite
Management of Change (MOC) category = RL
BAKR0800-05
0800-05
CAJT0800-5EA
755.37
CAD
CAJT0800-5
JT0800-5
Ammonium sulfate ≥99%, granular FCC, J.T.Baker®
Ammonium sulphate
Granular.
Formula:
(NH₄)₂SO₄ MW: 132.14 g/mol Melting Pt: 336…339 °C Density: 1.78 g/cm³ (20 °C) |
MDL Number:
MFCD00003391 CAS Number: 7783-20-2 Merck Index: 13,00559 |
Specification Test Results
Food GMP Manufactured Product | |
Meets F.C.C. Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Food Chemical | |
FCC - Assay | 99.0 - 100.5 % |
FCC - Identification | Passes Test |
FCC - Lead (Pb) | ≤3 mg/kg |
FCC - Residue on Ignition | ≤0.25 % |
FCC - Selenium (Se) | ≤0.003 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...