PolarSafe™ 100-Place Freezer Storage Boxes, Argos Technologies
Supplier: Antyila Scientific
24452-86 24452-87 24452-88 24452-89 24452-90 24452-91
89006-920CS 2186 CAD
89006-920 89006-922 89006-924 89006-926 89006-928 89006-930
PolarSafe™ 100-Place Freezer Storage Boxes, Argos Technologies
Boxes Cryoboxes
Storage boxes are suitable for use with 0.5, 1.5, and 2.0mL microtubes, as well as most other brands of cryogenic vials
Polypropylene boxes are a few millimeters taller than other available types of storage boxes and can accommodate several sizes of cryogenic vials or tubes of similar diameters up to 53.5mm (23/32") high. Boxes can withstand temperatures from –80° to 121°C (–112 to 250°F) and, unlike cardboard boxes, will not degrade after repeated thaws. The lid has an alphanumeric grid reference to aid with sample identification. Matte surfaces on box sides are suitable for marking orMore Product Information