Diammonium hydrogen orthophosphate ≥98% (by acidimetry), BAKER ANANYZED™ ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Synonyms:
Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate
Management of Change (MOC) category = NI
BAKR0784-07
BAKR0784-R
BAKR0784-01
BAKR0784-05
CAJT0784-7EA
2462.94
CAD
CAJT0784-7
CAJT0784-R
CAJT0784-1
CAJT0784-5
Diammonium hydrogen orthophosphate ≥98% (by acidimetry), BAKER ANANYZED™ ACS
di-Ammonium hydrogen orthophosphate
Crystals. Lot analysis on label.
Formula:
(NH₄)₂HPO₄ Melting Pt: 185 °C Density: 1.62 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010891 CAS Number: 7783-28-0 EINECS: 231-987-8 REACH: 01-2119490974-22 Merck Index: 13,00546 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay ((NH₄)₂HPO₄) (by acidimetry) | ≥98.0 % |
pH of 5% Solution at 25°C | 7.7 - 8.1 |
Insoluble Matter | ≤0.005 % |
Chloride (Cl) | ≤0.001 % |
Nitrate (NO₃) | ≤0.003 % |
Sulfate (SO₄) | ≤0.01 % |
Heavy Metals (as Pb) | ≤0.001 % |
Trace Impurities - Iron (Fe) | ≤0.001 % |
Calcium (Ca) | ≤0.001 % |
Magnesium (Mg) | ≤0.0005 % |
Potassium (K) | ≤0.005 % |
Sodium (Na) | ≤0.005 % |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...