KIMAX® Griffin Beakers, Low Form, Double Scale, Borosilicate Glass, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
14000-30 14000-1000 14000-2000 14000-10 14000-20 14000-100 14000-150 14000-250 14000-400 14000-600 14000-800 14000-1500 14000-4000 14000-50
89000-742CS 675.73 CAD
89000-742 89000-744 89000-746 89001-054 89001-056 89001-058 89001-060 89001-062 89001-064 89001-066 89001-068 89001-070 89001-072 89002-748
KIMAX® Griffin Beakers, Low Form, Double Scale, Borosilicate Glass, Kimble Chase
Beakers
These Griffin beakers feature thick, slightly flared, beaded tops with a spout designed for excellent pouring characteristics.
  • Durable matte finish marking area
  • White graduated scale for measuring and/or mixing where a high level of accuracy is not required
They have uniform walls throughout and are constructed of KIMAX KG-33 glass.
Certifications: Designed from ASTM Specification E960, Type I requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable