Ammonium carbonate 30,0% NH3 basis, lumps, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Salt of hartshorn

M.O.C. Category N

BAKR0642-01 BAKR0642-05
CAJT0642-1CS 919.07 CAD
CAJT0642-1 CAJT0642-5
Ammonium carbonate 30,0% NH3 basis, lumps, BAKER ANALYZED® ACS
Ammonium carbonate

Lot analysis on label.

Formula: (NH₄)₂CO₃
MW: 96.09 g/mol
Melting Pt: 58
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 506-87-6
EINECS: 208-058-0
Merck Index: 13,00508

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (as NH₃) ≥30.0 %
Insoluble Matter ≤0.005 %
Nonvolatile Matter ≤0.01 %
Sulfur Compounds (as SO₄) ≤0.002 %
Chloride (Cl) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤3.000 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR