Ammonium acetate ≥97%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Acetic acid ammonium salt
Management of Change (MOC) category = NI
J.T. Baker®
BAKR0596-R
0596-07
BAKR0596-07
BAKR0596-01
0596-01
0596-R
BAKR0596-05
0596-05
CAJT0596-REA
14788.66
CAD
JT0596-R
JT0596-01
JT0596-05
JT0596-7
CAJT0596-R
CAJT0596-1
CAJT0596-5
CAJT0596-7
Ammonium acetate ≥97%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Ammonium acetate
Formula:
H₃CCOONH₄ MW: 77.08 g/mol Melting Pt: 114 °C Density: 1.17 g/cm³ (20 °C) Flash Pt: 136 °C |
MDL Number:
MFCD00013066 CAS Number: 631-61-8 Merck Index: 13,00495 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (CH₃COONH₄) (by acid-base titration) | ≥97 % |
Insoluble Matter | ≤0.005 % |
Nitrate (NO₃) | ≤0.001 % |
pH of 5% Solution at 25°C | 6.7 - 7.3 |
Residue after Ignition | ≤0.010 % |
Solution (20:100) - clear, colorless, complete | Passes Test |
Sulfate (SO₄) | ≤0.001 % |
Water (H₂O) | ≤2.5 % |
Chloride (Cl) | ≤5 ppm |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤5 ppm |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...