Aluminium sulfate octadecahydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Trade termforalum, Aluminum sulfate octadecahydrate, Aluminum(III) sulfate octadecahydrate, Aluminum(III) sulphate octadecahydrate, Aluminum sulphate octadecahydrate

Management of Change (MOC) category = N

BAKR0564-01 0564-05
CAJT0564-1CS 1814.39 CAD
CAJT0564-1 CAJT0564-5 JT0564-5
Aluminium sulfate octadecahydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS
Aluminium sulphate octadecahydrate

Crystals. Lot analysis on label.

Formula: Al₂(SO₄)₃·18H₂O
Melting Pt: 86 °C
Density: 1.69 g/cm³ (20 °C)
MDL Number: MFCD00149136
CAS Number: 7784-31-8
EINECS: 233-135-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dace68759944aa1c6c0f56a372ea396f004e261&vid=e63de3c58650a97622a605c237aa52402ff157e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Al₂(SO₄)₃·18H₂O) (by EDTA titrn) 98.0 - 102.0 %
ACS - Insoluble Matter ≤0.005 %
ACS - Calcium (Ca) ≤0.01 %
ACS - Chloride (Cl) ≤0.005 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Iron (Fe) ≤0.002 %
ACS - Magnesium (Mg) ≤0.002 %
ACS - Potassium (K) ≤0.005 %
ACS - Sodium (Na) ≤0.02 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR