VITLAB® Griffin Beakers, PMP, BrandTech
Supplier: Brandtech
60503 60603 60803 60703 60903 61003 61103 61203 61403 61503 61603 61703 61803
83008-780PK 114.15 CAD
83008-780 83008-784 83008-792 83008-788 83008-796 83008-800 83008-804 83008-808 83008-816 83008-820 83008-824 83008-828 83008-832
VITLAB® Griffin Beakers, PMP, BrandTech
Beakers
High-quality beakers made of plastic polymethylpentene with moderately good chemical resistance.
  • Glass-clear polymethyl pentene (PMP)
  • Red screened graduations for high contrast with many liquids
Beakers feature molded graduations with red-screened overprint for legibility.
Certifications: All beakers meet the requirements of ISO 7056.