VITLAB® Low Form Griffin Beakers, PFA, BrandTech
Supplier: Brandtech
110205 110305 110405 110605 110905 111005
83008-622PK 125.44 CAD
83008-622 83008-626 83008-630 83008-634 83008-642 83008-646
VITLAB® Low Form Griffin Beakers, PFA, BrandTech
Beakers
Beakers are made of PFA (perfluoroalkoxy copolymer), a fluoropolymer with excellent chemical resistance
  • Containers provide excellent stability for standards and are manufactured without metal catalysts
  • Highly translucent for verification of contents
  • Thermally stable from –200 to 260°C (–328 to 500°F)
  • Molded graduations are easy-to-read and resistant to wear from repeated use
  • Autoclavable
Surfaces are much less porous than PTFE and are highly cleanable to reduce sample contamination by “memory effect” from prior samples. Ideal for use in all aspects ofMore Product Information