VWR®, Graduated Beakers with Handle, Low Form, Polypropylene
Supplier: VWR International
13916-707 13916-730 13916-762 13916-795 13916-820
13916-707CS 517.42 CAD
13916-707 13916-730 13916-762 13916-795 13916-820
VWR®, Graduated Beakers with Handle, Low Form, Polypropylene
Beakers
Graduated beakers are robust, with a non-drip spout and ridged thumb-grip on handle.
  • Molded of autoclavable, translucent polypropylene
  • Low form for better stability
  • All sizes graduated in milliliters and liters
Order Now
SPECIFICATIONS
Environmentally Preferable