Nalgene® Griffin Low Form Graduated Beakers, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
1201-0030 1201-0050 1201-0100 1201-0150 1201-0250 1201-0400 1201-0600 1201-1000 1201-2000 1201-4000
13915-067CS 327.22 CAD
13915-067 13915-078 13915-089 13915-090 13915-103 13915-114 13915-125 13915-147 13915-169 13915-180
Nalgene® Griffin Low Form Graduated Beakers, Polypropylene, Thermo Scientific
Beakers
Heavy-duty, translucent polypropylene beakers with ribs for stacking.
  • Easy-to-read graduations
  • Autoclavable
  • Chemically resistant
Beakers feature easy-to-read graduations with size and resin codes. Maximum use temperature and no-flame symbol printed in black. Autoclavable and chemically resistant. Not for use on a hot plate.