Nalgene® Graduated Griffin Beakers, PMP, Thermo Scientific
Supplier: Thermo Fisher Scientific
1203-0050 1203-0100 1203-0150 1203-0250 1203-0400 1203-0600 1203-1000 1203-2000 1203-4000
13918-043CS 375.87 CAD
13918-043 13918-065 13918-087 13918-101 13918-123 13918-145 13918-167 13918-189 13918-203
Nalgene® Graduated Griffin Beakers, PMP, Thermo Scientific
Beakers
Shatter-resistant PMP (polymethylpentene) has glass-like transparency, top heat tolerance and chemical resistance.
  • Non-toxic, non-contaminating, biologically inert
  • Safe for repeated autoclaving
  • Easy-to-read, silk-screened graduations, plus maximum temperature use, “no flame” symbol and resin code
  • Oval marking area, tapered walls
Takes constant usage at 150°C (302°F); intermittent at 175°C (347°F).
Certifications: Meet ISO/DIS 7056 international standards for laboratory plasticware accuracy.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO14001 (Certificate Registration No. EMS 752603)