VWR®, Griffin Beakers, Polypropylene
Supplier: VWR
VWR®
400300-0150 400300-0400 400300-0250 400300-4000 400300-1000 400300-0100 400300-0050 400300-2000 400300-0600
13890-160CS 812.86 CAD
13890-160 13890-181 13890-080 13890-104 13890-148 13890-126 13890-068 13890-046 13890-024
VWR®, Griffin Beakers, Polypropylene
Beakers
Translucent walls and molded graduations make it easy to prepare, mix, and measure solutions within the beaker.
  • Stackable
  • Sturdy construction withstands repeated autoclaving
  • Break-resistant, non-toxic, non-contaminating, and extremely chemical-resistant
  • Feature no-drip spout for clean pouring
  • Tapered walls for secure grip
  • Oval marking spot
Ideal beaker for general laboratory procedures.