PYREX® Berzelius Beakers, Tall Form, Graduated
Supplier: Corning
PYREX®
1060-400 1060-1L 1060-600 1060-100 1060-1L 1060-300 1060-200 1060-100 1060-500 1060-500
13922-029CS 565.58 CAD
13922-029 470211-362 13922-109 13922-084 13922-062 13922-040 13922-120 13922-142 470211-352 470211-354
PYREX® Berzelius Beakers, Tall Form, Graduated
Beakers
PYREX® Berzelius beakers are made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Autoclavable
Beakers with spout facilitates pouring. PYREX® Berzelius beakers are graduated to indicate their approximate content.
Certifications: Meets ASTM E960 specifications.