Mega Racks for 10 to 13 mm Tubes
Supplier: VWR International
77395-430 77395-432
77395-430CS 208.9 CAD
77395-430 77395-432
Mega Racks for 10 to 13 mm Tubes
Racks Tube Racks
Fully assembled, polypropylene mega racks are great for high throughput labs requiring large sample storage.
  • Sturdy, material for easy and secure transport of blood collection and other tubes
  • Alphanumeric identification for quick location and retrieval of tubes
  • Place different test groupings into one rack eliminating the need for multiple racks
The Mega Rack® are high-capacity storage racks.