Tube Racks, Coated Wire, 29 to 33 mm
Supplier: VWR International
77395-302 77395-304
77395-302CS 2350.79 CAD
77395-302 77395-304
Tube Racks, Coated Wire, 29 to 33 mm
Racks Tube Racks
Chemically resistant HDPE coated wire racks.
  • Holds all brands and types of conical, round or flat bottom, glass or plastic, tubes and/or bottles
  • Square wells and three deck levels help keep tubes secure
The HDPE coated wire rack is durable for transport or sample handling. It is chemically resistant and suitable for conical, round or flat-bottom, or plastic tubes and bottles.