Chemware® Griffin Beakers, PTFE, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
D1069035 D1069036 D1069037 D1069038 D1069039 D1069040 D1069041
13914-177EA 259.09 CAD
13914-177 13914-199 13914-213 13914-235 13914-257 13914-279 13914-290
Chemware® Griffin Beakers, PTFE, Saint-Gobain Performance Plastics
Beakers
These fluoropolymer beakers are non-contaminating and completely inert to chemical attack.
  • Unbreakable
  • Easy to clean
  • Lightweight
  • Microwavable
Beakers can be heated on a hot plate up to 288°C (550°F) when the temperature is monitored. Specially molded bottom allows rapid heat transfer. Broad spout for easy pouring.