Nalgene® Griffin Beakers, Low Form, Teflon® PFA, Thermo Scientific
Supplier: Thermo Fisher Scientific
1510-0050 1510-0100 1510-0250 1510-1000
13917-517CS 918.03 CAD
13917-517 13917-528 13917-540 13917-561 13917-572
Nalgene® Griffin Beakers, Low Form, Teflon® PFA, Thermo Scientific
Beakers
Beakers offer excellent chemical resistance for aggressive or high-purity chemicals and solutions.
  • Superior quality for tough lab use
  • Very wide temperature range: –270 to 250°C
  • Not for use on a hot plate
  • Autoclavable
Order Now
SPECIFICATIONS
Environmentally Preferable