Glass Jugs, Amber and Clear, KIMBLE® DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
5916438V-25 5916438B
89046-910CS 86.88 CAD
89046-910 89046-912 89046-918 89046-922 89046-924 89046-930 89046-938 89046-942 89046-920
Glass Jugs, Amber and Clear, KIMBLE® DWK Life Sciences
Bottles Carboys
Jugs are made of Type III soda-lime glass and feature narrow mouths and sturdy handles for safe pouring.
  • With or without black phenolic closures
Amber glass protects light-sensitive contents. Clear glass provides easy viewing of contents.
Ordering information: All jugs are packaged in corrugated cartons with divider cells and caps attached.