Nalgene® Validation Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
DS2127-0250 DS2127-2000 DS2127-0030
16172-153CS 274.29 CAD
16172-153 74910-034 74910-032
Nalgene® Validation Bottles, Thermo Scientific
Bottles Carboys
Ideal for QA lot sample storage of pharmaceuticals.
  • Tough, nontoxic containers are autoclavable for sterile applications
Bottles allow convenient material compatibility validation when scaling up production to larger containers. Constructed of clear polycarbonate and supplied with white polypropylene closures with TPE gasket. Bottles feature thick walls and wide mouths. Transparent design allows for visual inspection of contents.
Order Now
SPECIFICATIONS
Environmentally Preferable