SP Bel-Art Autoclavable Carboys, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F10794-0025 F10794-0050 F11846-0050 F11846-0025
47751-716EA 192.54 CAD
47751-716 47751-718 47751-722 74950-052
SP Bel-Art Autoclavable Carboys, Polypropylene, Bel-Art Products, a part of SP
Bottles Carboys
Rugged carboys for direct contact with foods and beverages, and are USP Class VI certified for the demands of the biotechnology and pharmaceutical industries.
  • Excellent chemical resistance
  • Large handles
  • Polypropylene construction
  • Autoclavable
  • Easy-to-use spigot
Ideal for storing and dispensing large batches of culture media, distilled water and other solutions, they have excellent chemical resistance and large handles that help with transport.
All components are FDA-grade polypropylene construction and are steam autoclavable at 121 °C (250 °F).More Product Information