Nalgene® Carboys with Tubulation, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
2301-0050 2301-0020
62200-100CS 1214.14 CAD
62200-100 62200-102
Nalgene® Carboys with Tubulation, Polypropylene, Thermo Scientific
Bottles Carboys
Carboys are ideal for a variety of dispensing applications.
  • Autoclavable
The tubulation can be connected to pumps, automated equipment, or filling bells with 1.27 cm (¹/₂") I.Ø. tubing (not included). The tubulation is molded into one piece with the bottle and positioned low on the carboy for maximum drainage. It is graduated to deliver in 5 L and 1 gal. increments.
They feature an outlet positioned low on the carboy for efficient drainage as well as conveniently placed wide shoulder handles for easy transport.
Ordering information: Supplied with 83B polypropylene closures.