VWR® StaticAway™ Antistatic Brush
Supplier: VWR International
89094-716 89094-718 89094-720 15551-016
89094-716EA 59.9 CAD
89094-716 89094-718 89094-720 15551-016
VWR® StaticAway™ Antistatic Brush
Brushes
Brushes clean by dissipating static and whisking away harmful dust, lint, and dirt.
  • Provide accurate results by eliminating pan weight variances caused by static
  • Long-handled brushes allow access to difficult places
Ideal for cleaning balance pans, optics, cuvettes, slides, film, microscopes, electronics, and computer monitors. Bristles will not scratch critical surfaces. Brushes drain static and clean with synthetic conductive fibers and a conductive composite handle. The compact brush is suitable for electronic balances, providing accurate resultsMore Product Information