Narrow Tubing Brushes, Mill-Rose
Supplier: Mill-Rose
72083W 72084W
17070-000PK 22.21 CAD
17070-000 17130-069
Narrow Tubing Brushes, Mill-Rose
Brushes
Feature short, close bristles for effective cleaning of narrow tubing, burets, pipets, and funnel stems.
Natural white bristles on twisted wire handle with a convenient loop for hanging.