Centrifuge Tube Brushes, Mill-Rose
Supplier: Mill-Rose
72070W 72072W 72071W
17130-044PK 101.31 CAD
17130-044 17130-066 17130-022
Centrifuge Tube Brushes, Mill-Rose
Brushes
Tapered natural bristle construction for cleaning centrifuge tubes, vials, and small funnels.
  • Flexible twisted wire handle has a convenient hanging loop
  • Select models available with black nylon tufted ends
  • Fan tip model provides effective cleaning for conical tubes