Nalgene® Dispensing and Storage Jugs, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
2221-0010 2221-0020
16093-011CS 556.16 CAD
16093-011 16093-022
Nalgene® Dispensing and Storage Jugs, Polypropylene, Thermo Scientific
Bottles Carboys
Autoclavable, leakproof jugs offer excellent chemical resistance.
  • Leakproof
  • Autoclavable
Note: Before autoclaving, set cap on top of jug without engaging the threads.
A large carrying handle allows for the use of rubber gloves for safe handling of corrosive liquids, as well as an offset pour spout for easy, accurate pouring.
Ordering information: Jugs are supplied with polypropylene screw closures. For replacement caps, see 16219-122 series
Order Now
SPECIFICATIONS
Environmentally Preferable