Beaker and Jar Brushes, Mill-Rose
Supplier: Mill-Rose
MR-75002W MR-75001B MR-75010W
17176-023EA 134 CAD
17176-023 17180-009 17181-001
Beaker and Jar Brushes, Mill-Rose
Brushes
Designed for heavy-duty cleaning of beakers, jars, and other wide-mouth glassware.
  • Heavy tufting at the tip reduces scratching and improves cleaning capability
  • Handle has an opening for hanging
Long-lasting bristles are spirally wound into grooves in the smooth wood handle.
Order Now
SPECIFICATIONS
Environmentally Preferable