Nalgene® Rectangular Carboys with Handle, High-Density Polyethylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
2211-0020 2211-0050
16115-012CS 973.94 CAD
16115-012 16115-034
Nalgene® Rectangular Carboys with Handle, High-Density Polyethylene, Thermo Scientific
Bottles Carboys
Sturdy, space-saving design with molded-in graduations in liters and gallons.
  • Leakproof
For replacement caps, see cap 16219-106 in 16219-030 series.
Built-in shoulder loops have convenient stainless steel bail handle attached. Wide-mouth opening for easy filling, transferring, and cleaning. Supplied with cap. Cap size: 100-415.
Order Now
SPECIFICATIONS
Environmentally Preferable