Nalgene® Rectangular Carboys with Spigot and Handle, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
2321-0020 2321-0050
16115-191CS 1852.52 CAD
16115-191 16115-216
Nalgene® Rectangular Carboys with Spigot and Handle, Polypropylene, Thermo Scientific
Bottles Carboys
For dispensing solutions, media, and sterile water.
  • 8.9 cm (3¹/₂") wide mouth for easy filling and cleaning
  • Sturdy handles
  • Graduated in liters and gallons
  • Leakproof
  • Autoclavable
Note: Autoclave only when empty, using a properly vented closure. Dismount and disassemble spigot before autoclaving.
Ordering information: Supplied with carboy, polypropylene spigot, and 100-415 screw closure. For replacement caps, see cap 16219-106 in 16219-030 series.