Nalgene® Microcentrifuge Tube Boxes, Polycarbonate, Thermo Scientific
Supplier: Thermo Fisher Scientific
5055-5002 5055-5005 5055-5015
55710-184CS 934.42 CAD
55710-184 55710-173 55710-175
Nalgene® Microcentrifuge Tube Boxes, Polycarbonate, Thermo Scientific
Boxes Cryoboxes
Provide safe, secure storage of filled microcentrifuge tubes from –150°C to 121°C (not for use in liquid-phase nitrogen)
High-temperature permanent foam insert allows repeated autoclaving. Transparent polycarbonate lid has printed grid for inventory purposes, permits easy viewing of contents, is removable for easy access, and is keyed to orient front and back of box. Lid accepts writing with marker for box and tube identification.