PureCube IDA XL Agarose

55603 55605 55610 55612
77393-276EA 250.62 CAD
77393-276 77393-278 77393-280 77393-282
PureCube IDA XL Agarose
Protein Purification Protein Purification Reagents

PureCube IDA XL (iminodiacetic acid) agarose resin is used for IMAC-based protein purification and protein enrichment methods.


  • Usage: For coupling of metal ions for corresponding purification and/or enrichment purposes
  • Specificity: Varies
  • Binding capacity: Depends on the loaded ion
  • Purity: Depends on the loaded ion
  • Bead Ligand: IDA
  • Bead size: 400 μm
  • Matrix: 7.5% cross-linked agarose
  • Filling quantity: Delivered as a 50% suspension
  • Recommended FPLC flow rates: 0.5 to 2 ml/min
  • Maximal FPLC flow rate: 6 ml/min
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile


The beads in this suspension have an average diameter of 400 µm, which is very large. This size is useful for very high flow rates in, e.g., FPLC assays or very viscous cell media. Since the affinity of an IDA-metal-ion-complex depends on the loaded metal ion, we offer IDA agarose resin uncoupled to any metal ion in case we do not offer your desired IDA + metal ion combination. Through our coupling protocol, you can, in turn, attach your desired metal ions to the agarose beads.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR