Nalgene® Rectangular Carboys with Handle, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
2212-0020 2212-0050
16115-125CS 1437.12 CAD
16115-125 16115-147
Nalgene® Rectangular Carboys with Handle, Polypropylene, Thermo Scientific
Bottles Carboys
Ideal for storing solutions, media, and sterile water.
  • Leakproof
For replacement caps, see cap 16219-106 in 16219-030 series.
Note: Before autoclaving, set cap on top of carboy without engaging threads.
Useful for handling large volumes of powders and other solid samples. Wide mouth allows easy, efficient cleaning. Space-saving carboys molded in tough, autoclavable polypropylene with 8.9 cm (3¹/₂") neck opening. Sturdy handle attached to molded-in shoulder loops. Calibrated in liters and gallons. Cap size: 100-415.