Ammonium sulfate ≥99.0%, GR ACS, Supelco®

Supplier: MilliporeSigma

AX1385-3 AX1385-5
CAAX1385-1EA 156.83 CAD
CAAX1385-1 CAAX1385-3 CAAX1385-5
Ammonium sulfate ≥99.0%, GR ACS, Supelco®
Ammonium sulphate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: (NH₄)₂SO₄
Melting Pt: >280 °C (dec., lit.)
Density: 1.77 g/ml (25°C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Specification Test Results

Assay 99.0% min
Chloride (Cl) 5ppm max
Color White
Form Crystals
Heavy metals (as Pb) 5ppm max
Insoluble matter 0.005% max
Iron (Fe) 5ppm max
Nitrate (NO3) 0.001% max
pH of a 5% solution at 25°C 5.0-6.0
Phosphate (PO4) 5ppm max
Residue after ignition 0.005% max

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR