PureCube Compact Cartridge Al-NTA

77393-630EA 304.32 CAD
77393-630 77393-638 77393-614 77393-622
PureCube Compact Cartridge Al-NTA
Protein Purification Protein Purification Reagents

PureCube Al-NTA compact cartridge is an FPLC-ready column pre-packed with our PureCube Al-NTA agarose resin. It is used for the purification of His-tagged proteins and the enrichment of phosphorylated proteins.


  • Usage:Enrichment of phosphopeptides, specific binding and purification of 6× His-tagged proteins
  • Specificity: Affinity to phosphorylated biomolecules
  • Bead ligand: Al-NTA
  • Bead size: 40 µm
  • Filling quantity: Delivered as a 50% suspension
  • Matrix: 7.5% crosslinked agarose
  • Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile


As an alternative, we also offer PureCube Al-NTA MagBeads, the more common format of Fe-NTA beads for phosphopeptide enrichment.


The PureCube cartridge/column that is used here is compatible with almost all commonly used FPLC systems like ÄKTA TM.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR