Adson Tissue Forceps, Sklar
Supplier: Sklar
47-1447 47-2047 47-2049 47-2147 50-3047 50-3048 50-3049
82030-250EA 163.91 CAD
82030-250 82030-252 82030-254 82030-256 82030-272 82030-274 82030-276
Adson Tissue Forceps, Sklar
Forceps
Adson Tissue Forceps are autoclavable and are made from premium-grade stainless steel. And Censitrac ready products are industry standard and constructed of stainless steel.
Adson-Brown Tissue Forceps and Tissue Forceps are designed for holding and manipulating delicate tissues
Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They have a wide thumb grasp for increased precision and control.
These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.