Acetonitrile, anhydrous ≥99.9% (by GC) for biotechnology, Supelco®

Supplier: MilliporeSigma
Danger

AX0152-NP20 AX0152-1 AX0152-2505
CAAX0152-NP20EA 2449 CAD
CAAX0152-NP20 CAAX0152-1 CAAX0152-2505
Acetonitrile, anhydrous ≥99.9% (by GC) for biotechnology, Supelco®
Acetonitrile

For use in DNA synthesis. Flammable. Toxic.

Formula: H₃CCN
Boiling Pt: 81…82 °C (lit.)
Melting Pt: –45 °C (lit.)
Density: 0.786 g/ml (25°C) (lit.)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.9% min
Appearance Clear liquid, free from particulates
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Free amine (as dimethylamine) 1ppm max
Residue after evaporation 5ppm max
Titrable acid 2.0µeq/g max
Titrable base 0.6µeq/g max
UV Abs. at 200 nm 1.00AU max
UV Abs. at 210 nm 0.60AU max
UV Abs. at 220 nm 0.30AU max
UV Abs. at 230 nm 0.08AU max
UV Abs. at 240 nm 0.03AU max
UV Abs. at 250 nm 0.03AU max
UV Abs. at 270 nm 0.02AU max
UV Abs. at 300 nm 0.01AU max
UV Abs. at 350 nm 0.01AU max
UV Cut-off 200nm max
Water (H2O) 10ppm max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR