Acetonitrile ≥99.5% (by GC), GR ACS, Supelco®
Supplier: MilliporeSigma
|
Danger
|
AX0149-20
AX0149-3
AX0149-1
AX0149-6
AX0149-1
CAAX0149-20EA
6368
CAD
CAAX0149-20
CAAX0149-3
CAAX0149-1
CAAX0149-6
EM-AX0149-1
Acetonitrile ≥99.5% (by GC), GR ACS, Supelco®
Acetonitrile
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
Formula:
H₃CCN Boiling Pt: 81…82 °C (lit.) Melting Pt: –45 °C (lit.) Density: 0.786 g/ml (25°C) (lit.) Flash Pt: 2 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II REACH: 01-2119471307-38 Merck Index: 13,00071 |
|
Specification Test Results
Assay (GC) | 99.5% min |
Appearance | Clear |
Color (APHA) | 10 max |
Residue after evaporation | 0.005% max |
Titrable acid | 8µeq/g max |
Titrable base | 0.6µeq/g max |
Water (H2O) | 0.3% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...