Acetonitrile ≥99.8% for HPLC, Supelco®
Supplier: MilliporeSigma
|
Danger
|
AX0145-NP20
AX0145P-6
AX0145-1
AX0145P-1
CAAX0145-36EA
18823.47
CAD
CAAX0145-36
CAAX0145-37
CAAX0145-38
CAAX0145-40
CAAX0145-NP20
CA-AX0145P-6
CAAX0145-1
CAAX0145P-1
Acetonitrile ≥99.8% for HPLC, Supelco®
Acetonitrile
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
Formula:
H₃CCN Boiling Pt: 81.6 °C (1013 hPa) Melting Pt: –45.7 °C Density: 0.782 g/cm³ (20 °C) Flash Pt: 2 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II REACH: 01-2119471307-38 Merck Index: 13,00071 |
|
Specification Test Results
Appearance | Clear liquid, free from particulates |
Assay (GC) | 99.8% min |
Color (APHA) | 10 max |
Residue after evaporation | 1ppm max |
Filtered through 0.2 µm filter | To pass test |
Gradient at 254 nm | 0.005AU max |
Infrared spectrum | Conforms to standard |
Titrable acid | 0.008meq/g max |
Titrable base | 0.0006meq/g max |
UV Abs. at 190 nm | 1.00AU max |
UV Abs. at 220 nm | 0.05AU max |
UV Abs. at 254 - 400 nm | 0.01AU max |
UV Cut-off | 190nm max |
Water (H2O) | 0.10% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...