Zinc oxide ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
11137-09
11137-18
11137-30
CAAA11137-09EA
103.96
CAD
CAAA11137-09
CAAA11137-18
CAAA11137-30
Zinc oxide ≥99.999% (metals basis), Puratronic®
Zinc oxide
Powder. Purity based on metal contaminates only.
Purity: 99.999%
CAS No.: 1314-13-2
Molecular Formula: ZnO
Formula Weight: 81.37
Physical Form: Powder
Density: 5.606
Melting Point: 1975°
Merck Reference: 14,10147
UN No.: 3077
MDL No.: MFCD00011300
Formula:
ZnO Melting Pt: 1975 °C Density: 5.606 Storage Temperature: Ambient |
MDL Number:
MFCD00011300 CAS Number: 1314-13-2 EINECS: 215-222-5 UN: 3077 ADR: 9,III REACH: 01-2119463881-32 Merck Index: 14,10147 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...