Nalgene® Rectangular Carboys with Spigot and Handle, Polycarbonate, Thermo Scientific
Supplier: Thermo Fisher Scientific
2322-0020 2322-0050
16115-373CS 2504.6 CAD
16115-373 16115-395
Nalgene® Rectangular Carboys with Spigot and Handle, Polycarbonate, Thermo Scientific
Bottles Carboys
Transparent carboy features molded-in graduations
  • Leakproof
For replacement caps, see cap 16219-106 in 16219-030 series.
Note: Before autoclaving, set cap on top of carboy without engaging threads and disassemble spigot.
High-impact strength enables reliable cold room use. Wide mouth allows easy, efficient cleaning. Autoclave only when empty. Cap size: 100-415.
Ordering information: Supplied with carboy, polypropylene screw cap, and spigot.
Order Now
SPECIFICATIONS
Environmentally Preferable