Beaker, Jar, and Cylinder Brushes, Mill-Rose
Supplier: Mill-Rose
72110B 72112B 2111B
17190-002EA 76.96 CAD
17190-002 17170-005 17190-007
Beaker, Jar, and Cylinder Brushes, Mill-Rose
Brushes
Completely rounded tip provides safe, effective cleaning for wide mouth glassware such as beakers, jars, and cylinders.
  • Extra dense natural black bristles are mounted on twisted wires into a durable wood handle
  • Double-tuft extensions at the tip for reaching into corners
  • Available with two or four rows of bristles