Acetone ≥99.5% (by GC, corrected for water content) for HPLC, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
2-Propanone, DMK, Propanone, Dimethyl ketone
AX0115-20
AX0115-6
AX0115-NP20
AX0115-1
AX0115P-1
AX0115P-6
CAAX0115-20EA
5733
CAD
CAAX0115-20
CAAX0115-38
CAAX0115-39
CAAX0115-40
CAAX0115-6
CAAX0115-NP20
CAAX0115-1
CAAX0115P-1
CAAX0115P-6
Acetone ≥99.5% (by GC, corrected for water content) for HPLC, Supelco®
Acetone
HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs.
Formula:
CH₃COCH₃ Boiling Pt: 56.2 °C (1013 hPa) Melting Pt: –95.4 °C Density: 0.792 g/cm³ (20 °C) Flash Pt: <–20 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3,II REACH: 01-2119471330-49 Merck Index: 13,00067 |
|
Specification Test Results
Assay (GC, corrected for water content) | 99.5% min |
Aldehyde (as HCHO) | 0.002% max |
Appearance | Clear liquid, free from particulates |
Color (APHA) | 10 max |
Filtered through 0.2 µm filter | To pass test |
Form | Liquid |
Infrared spectrum | Conforms to standard |
Isopropyl alcohol | 0.05% max |
Methanol | 0.05% max |
Residue after evaporation | 5ppm max |
Solubility in water | To pass test |
Substances reducing permanganate | To pass test |
Titrable acid | 0.0003meq/g max |
Titrable base | 0.0006meq/g max |
UV Abs. at 330 nm | 1.00AU max |
UV Abs. at 340 nm | 0.10AU max |
UV Abs. at 350 nm | 0.02AU max |
UV Abs. at 400 nm | 0.01AU max |
UV Cut-off | 330nm max |
Water (H2O) | 0.5% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...