Acetic acid glacial ≥99%, OmniTrace® for trace metal analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ100
AX0077-1
AX0077-2
CAAX0077-1EA
209
CAD
CAAX0077-1
CAAX0077-2
Acetic acid glacial ≥99%, OmniTrace® for trace metal analysis, Supelco®
Acetic acid
For elemental metal analysis via atomic absorption and ICP instruments. Actual Lot Analysis on label. Color-coded label to match acid cap.
Formula:
H₃CCOOH Storage Temperature: Ambient |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 EINECS: 200-580-7 UN: 2789 ADR: 8,II |
|
Specification Test Results
Aluminum (Al) | 1ppb max |
Antimony (Sb) | 0.5ppb max |
Arsenic (As) | 0.5ppb max |
Assay | 99% min |
Barium (Ba) | 0.5ppb max |
Beryllium (Be) | 0.1ppb max |
Bismuth (Bi) | 0.1ppb max |
Cadmium (Cd) | 0.5ppb max |
Calcium (Ca) | 1ppb max |
Cerium (Ce) | 0.1ppb max |
Cesium (Cs) | 0.1ppb max |
Chloride (Cl) | 1ppm max |
Chromium (Cr) | 1ppb max |
Cobalt (Co) | 0.1ppb max |
Color (APHA) | 10 max |
Copper (Cu) | 0.5ppb max |
Dysprosium (Dy) | 0.1ppb max |
Erbium (Er) | 0.1ppb max |
Europium (Eu) | 0.1ppb max |
Gadolinium (Gd) | 0.1ppb max |
Gallium (Ga) | 0.1ppb max |
Germanium (Ge) | 0.5ppb max |
Hafnium (Hf) | 0.1ppb max |
Holmium (Ho) | 0.1ppb max |
Indium (In) | 0.1ppb max |
Iron (Fe) | 1ppb max |
Lanthanum (La) | 0.1ppb max |
Lead (Pb) | 0.1ppb max |
Lithium (Li) | 0.1ppb max |
Lutetium (Lu) | 0.1ppb max |
Magnesium (Mg) | 0.5ppb max |
Manganese (Mn) | 0.5ppb max |
Mercury (Hg) | 1ppb max |
Molybdenum (Mo) | 0.5ppb max |
Neodymium (Nd) | 0.1ppb max |
Nickel (Ni) | 0.5ppb max |
Phosphate (PO4) | 1ppm max |
Platinum (Pt) | 0.5ppb max |
Potassium (K) | 1ppb max |
Praseodymium (Pr) | 0.1ppb max |
Rhenium (Re) | 0.1ppb max |
Rhodium (Rh) | 0.5ppb max |
Rubidium (Rb) | 0.1ppb max |
Ruthenium (Ru) | 0.5ppb max |
Samarium (Sm) | 0.1ppb max |
Scandium (Sc) | 0.1ppb max |
Selenium (Se) | 1ppb max |
Silver (Ag) | 1ppb max |
Sodium (N/A) | 1ppb max |
Strontium (Sr) | 0.5ppb max |
Substances reducing dichromate | To Pass Test |
Substances reducing permangaN/Ate | To Pass Test |
Sulfate (SO4) | 0.5ppm max |
Tellurium (Te) | 0.5ppb max |
Terbium (Tb) | 0.1ppb max |
Thallium (Tl) | 0.1ppb max |
Thorium (Th) | 0.1ppb max |
Thulium (Tm) | 0.1ppb max |
Tin (Sn) | 0.5ppb max |
Titanium (Ti) | 0.5ppb max |
Tungsten (W) | 0.5ppb max |
Uranium (U) | 0.1ppb max |
VaN/Adium (V) | 0.5ppb max |
Ytterbium (Yb) | 0.1ppb max |
Yttrium (Y) | 0.1ppb max |
Zinc (Zn) | 1ppb max |
Zirconium (Zr) | 0.1ppb max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...