Acetic acid glacial ≥99.7%, GR ACS ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

MQ200

AX0073-21 AX0073-59 AX0073-6 AX0073-75 AX0073-78 AX0073-9 AX0073P-5
CAAX0073-21EA 3708 CAD
CAAX0073-21 CAAX0073-59 CAAX0073-6 CAAX0073-75 CAAX0073-78 CAAX0073-9 CAAX0073P-5
Acetic acid glacial ≥99.7%, GR ACS ACS, Supelco®
Acetic acid

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118 (lit.)
Melting Pt: 17 (lit.)
Density: 1.049 g/ml (25°C) (lit.)
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8.3 II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR