Rochester-Ochsner Hemostatic Forceps, Sklar
Supplier: Sklar
17-2655 17-2662 17-2672 17-2680 17-2872 17-2880
82029-996EA 120.83 CAD
82029-996 82029-998 82030-000 82030-002 82030-008 82030-010
Rochester-Ochsner Hemostatic Forceps, Sklar
Forceps
Rochester-Oschner (Kocher) Forceps are a heavy instrument designed to aggressively grasp medium to heavy tissue or occlude heavy, dense vessels.
  • Autoclavable and latex-free
  • Available either curved or straight, in multiple lengths
  • Forceps have horizontal serrations the entire length of the jaw
  • Serrations plus teeth ensure a firm grip on the tissue or vessel
These forceps are sometimes referred to as simply Oschner Forceps, however, Rochester-Oschner Forceps are actually heavier than standard Oschner Forceps.
Censitrac Ready OR Grade Rochester-Ochsner Forceps are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.