Nalgene® Carboys with Handles, Wide Mouth, Low-Density Polyethylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
2234-0020 2234-0030 2234-0050
16133-025CS 939.31 CAD
16133-025 16133-047 16133-058
Nalgene® Carboys with Handles, Wide Mouth, Low-Density Polyethylene, Thermo Scientific
Bottles Carboys
For corrosive liquids and solids.
  • Leakproof
For replacement caps, see cap 16219-106 in 16219-030 series.
Wide mouth allows easy access for cleaning. Convenient shoulder handles. Graduated to contain in 1 gal. and 5 L increments. Cap Size: 100-415.