Nalgene® Carboys with Handles, Low-Density Polyethylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
2210-0130 2210-0020 2210-0040 2210-0050 2210-0065
16117-062EA 454.42 CAD
16117-062 16117-007 16117-018 16117-029 16117-040
Nalgene® Carboys with Handles, Low-Density Polyethylene, Thermo Scientific
Bottles Carboys
Useful for storing and containment of chemicals and standards.
  • Wide shoulder handles for easy carrying and pouring
  • Supplied with ca
  • Cap size: 83B
  • Accepts No. 13¹/₂ rubber stopper
  • Leakproof
Ordering information: For replacement caps, see cap 16225-234 in 16219-122 series.
Order Now
SPECIFICATIONS
Environmentally Preferable