Dressing Forceps, Delicate, Sklar
Supplier: Sklar
22-1161 22-1162 22-1163 22-1164
89025-220EA 76.33 CAD
89025-220 89025-222 89025-224 89025-226
Dressing Forceps, Delicate, Sklar
Forceps
Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds
Censitrac Ready OR Grade Dressing Forceps are industry standard and constructed of stainless steel.
This product is straight with serrated tips.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.