Reagent Reservoir Chillers, 50/55 ml
Supplier: Heathrow Scientific
120730 120731 120732
77395-284CS 1650.88 CAD
77395-284 77395-286 77395-288
Reagent Reservoir Chillers, 50/55 ml
Reagent Reservoirs
Ice-free, non-toxic method to extend the working period and preserve integrity for temperature-sensitive reagents.
  • Reagents are kept at less than 5 °C (41 °F) for approximately 1 hour
  • Eliminates the risk of contamination from messy ice baths
  • Not for use with liquid nitrogen
  • Symmetrical cooler design allows reagent reservoirs to be mounted in either direction
  • Stackable design saves storage and work space
  • 2-year warranty
Engineered custom fit design for Heathrow Scientific 50 ml or 100 ml reservoirs to ensure a maximum contact for longer uniform cooling.