Reagent Reservoirs, 50 ml
Supplier: Heathrow Scientific
120646 120643 120644 120645
77395-266CS 1078.06 CAD
77395-266 77395-260 77395-262 77395-264
Reagent Reservoirs, 50 ml
Reagent Reservoirs
Reagent Reservoirs by Heathrow Scientific offer a fresh and modern design with enhanced features to streamline pipetting procedures.
  • Superior durability with enhanced walls and crush-resistant 'turrets' at the corners
  • Easy handling with tabs to prevent stacked reservoirs from sticking together
  • Graduation markings on the top edge and inner walls allow measurements of liquid to be easily identified and reduce reagent waste
  • Pour-off spouts conveniently located on all four corners for easy spill-free pouring and reduced dripping
  • Divided reservoir design allows
More Product Information